1,3-Dibromo-2-iodobenzene 97%
Supplier: Ambeed
|
Warning
|
A183262-1G
A183262-25G
A183262-5G
A183262-250MG
103815-626EA
27
USD
103815-626
103846-188
77083-784
77083-786
77083-788
77151-684
1,3-Dibromo-2-iodobenzene 97%
1,3-Dibromo-2-iodobenzene
Precaución: for Research Use Only
Formula:
C₆H₃Br₂I Boiling Pt: 303.7±22.0 °C (760 mm Hg) Melting Pt: 99.8 °C Flash Pt: 137.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00192666 CAS Number: 19821-80-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...