1,3-Dibromo-2-fluorobenzene 97%

Supplier: Ambeed
Warning

A217281-100G A217281-100MG A217281-10G A217281-250MG A217281-1G A217281-25G A217281-5G
103818-614EA 11 USD
103818-614 103849-346 76873-840 76873-842 76873-844 76873-846 77087-962 77087-964 77087-966
1,3-Dibromo-2-fluorobenzene 97%
1,3-Dibromo-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₃Br₂F
Storage Temperature: Ambient
MDL Number: MFCD13194294
CAS Number: 1435-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR