1,3-Dibromo-2-fluoro-5-nitrobenzene 97%

Supplier: Ambeed
Warning

A111774-100MG A111774-10G A111774-250MG A111774-25G A111774-1G A111774-5G
103797-902EA 15 USD
103797-902 103823-722 76809-960 76809-962 76809-964 76809-966 77310-532 77310-534
1,3-Dibromo-2-fluoro-5-nitrobenzene 97%
1,3-Dibromo-2-fluoro-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂FNO₂
Boiling Pt: 289.2 °C (760 mm Hg)
Flash Pt: 128.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07779294
CAS Number: 361436-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR