1,3-Dibromo-2-chlorobenzene 96%

Supplier: Ambeed
Warning

A306366-1G A306366-25G A306366-5G A306366-100G A306366-10G A306366-250MG A306366-500G
103814-462EA 6 USD
103814-462 103835-628 76838-328 76838-330 76838-332 76977-814 76977-816 76977-818 76977-820
1,3-Dibromo-2-chlorobenzene 96%
1,3-Dibromo-2-chlorobenzene

Precaución: For research use only

Formula: C₆H₃Br₂Cl
MW: 270.3500 g/mol
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD12922600
CAS Number: 19230-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR