N,N'-Bis(tert-butoxycarbonyl)-S-methylisothiourea 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl N,N'-Bis(tert-butoxycarbonyl)carbamimidothioate, N,N'-Di-Boc-carbamimidothioic Acid Methyl Ester, 1,3-Di-Boc-2-methylisothiourea, N,N'-Bis(tert-butoxycarbonyl)carbamimidothioic Acid Methyl Ester, N,N'-Di-Boc-S-methylisothiourea, Methyl N,N'-Di-Boc-carbamimidothioate, 1,3-Bis(tert-butoxycarbonyl)-2-methyl-2-thiopseudourea
A240222-25G
A240222-5G
A240222-100G
A240222-10G
A240222-500G
A240222-1G
103811-340EA
7
USD
103811-340
103851-138
77090-288
77090-290
77193-302
77193-304
77193-306
77304-190
N,N'-Bis(tert-butoxycarbonyl)-S-methylisothiourea 97%
N,N'-Bis(tert-butoxycarbonyl)-S-methylisothiourea
Precaución: for Research Use Only
Formula:
C₁₂H₂₂N₂O₄S Melting Pt: 115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00239356 CAS Number: 107819-90-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...