1,3-Di-boc-2-(trifluoromethylsulfonyl)guanidine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,3-Bis(tert-butoxycarbonyl)-2-(trifluoromethanesulphonyl)guanidine, Goodman's Reagent, 1,3-Bis(tert-butoxycarbonyl)-2-(trifluoromethylsulphonyl)guanidine, 1,3-Di-Boc-2-(trifluoromethylsulphonyl)guanidine, 1,3-Di-Boc-2-(trifluoromethanesulphonyl)guanidine
A144671-10G
A144671-250MG
A144671-1G
A144671-25G
A144671-5G
103803-436EA
30
USD
103803-436
103841-512
77190-566
77190-568
77316-762
77316-764
77316-766
1,3-Di-boc-2-(trifluoromethylsulfonyl)guanidine 95%
1,3-Di-boc-2-(trifluoromethylsulfonyl)guanidine
Precaución: for Research Use Only
Formula:
C₁₂H₂₀F₃N₃O₆S MW: 391.36 g/mol Melting Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD02683528 CAS Number: 207857-15-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...