1,3-Di(pyridin-4-yl)benzene 97%

Supplier: Ambeed
Warning

A212717-100MG A212717-250MG
103818-228EA 100 USD
103818-228 103848-924 77237-430 77746-289
1,3-Di(pyridin-4-yl)benzene 97%
1,3-Di(pyridin-4-yl)benzene

Precaución: for Research Use Only

Formula: C₁₆H₁₂N₂
MW: 232.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30478638
CAS Number: 170165-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR