1,3-Bis(hydroxymethyl)-5,5-dimethylimidazolidine-2,4-dione 95%
Supplier: Ambeed
|
Warning
|
A379332-5G
A379332-100G
A379332-10G
A379332-1G
A379332-25G
103837-676EA
13
USD
103837-676
76840-986
77195-606
77195-608
77195-610
77306-910
1,3-Bis(hydroxymethyl)-5,5-dimethylimidazolidine-2,4-dione 95%
1,3-Bis(hydroxymethyl)-5,5-dimethylimidazolidine-2,4-dione
Precaución: for Research Use Only
Formula:
C₇H₁₂N₂O₄ Boiling Pt: 303.7 °C (760 mm Hg) Melting Pt: 86…88 °C Flash Pt: 137.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00467199 CAS Number: 6440-58-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...