1,3-Bis(diphenylphosphino)propanenickel(II) chloride 98%

Supplier: Ambeed
Danger

Synonyms: [1,3-Bis(diphenylphosphino)propane]nickel (II) chloride, Dichloro[bis(1,3-diphenylphosphino)propane]nickel(II), 1,3-Bis(diphenylphosphino)propane nickel(II)chloride, Dichloro[1,3-bis(diphenylphosphino)propane]nickel(II), [1,3-Bis(diphenylphosphino)propane]dichloronickel(II), [1,3-Bis(diphenylphosphino)propane]nickel(II) dichloride

A133473-100G A133473-10G A133473-500G A133473-5G A133473-25G
103808-460EA 276 USD
103808-460 76996-518 76996-520 76996-522 77298-634 77314-530
1,3-Bis(diphenylphosphino)propanenickel(II) chloride 98%
1,3-Bis(diphenylphosphino)propanenickel(II) chloride

Precaución: for Research Use Only

Formula: C₂₇H₂₆Cl₂NiP₂
MW: 542.042 g/mol
Melting Pt: 213 °C
Flash Pt: 291.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00015318
CAS Number: 15629-92-2
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR