1,3-Bis(diphenylphosphino)propane 97%

Supplier: Ambeed
Warning

Synonyms: Propylenebis(diphenylphosphine), Propane-1,3-diylbis(diphenylphosphane), Propane-1,3-diylbis(diphenylphosphine), 1,3-Bis(diphenylphosphino)propane, DPPP, Bis(1,2-diphenylphosphino)propane

A150622-1000G A150622-100G A150622-10G A150622-500G A150622-25G A150622-5G
103842-326EA 5 USD
103842-326 76812-176 76812-178 76812-180 76812-182 77317-856 77317-858
1,3-Bis(diphenylphosphino)propane 97%
1,3-Bis(diphenylphosphino)propane

Precaución: for Research Use Only

Formula: C₂₇H₂₆P₂
Boiling Pt: 529.7 °C (760 mm Hg)
Melting Pt: 63…65 °C
Flash Pt: 235 °C
Storage Temperature: Ambient
MDL Number: MFCD00003050
CAS Number: 6737-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR