1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride 97%

Supplier: Ambeed
Warning

A214416-1G A214416-25G A214416-5G A214416-100G A214416-10G A214416-250MG
103818-400EA 12 USD
103818-400 103849-106 77087-624 77087-626 77087-628 77192-756 77192-758 77192-760
1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride 97%
1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride

Precaución: for Research Use Only

Formula: C₂₇H₃₇ClN₂
Melting Pt: 278 °C
Storage Temperature: Ambient
MDL Number: MFCD02684545
CAS Number: 250285-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR