1,3-Benzothiazole-2-carbaldehyde 98%

Supplier: Ambeed
Danger

A688982-10G A688982-250MG A688982-1G A688982-5G A688982-100G
103840-568EA 73 USD
103840-568 76651-078 76651-080 77308-118 77308-120 77749-418
1,3-Benzothiazole-2-carbaldehyde 98%
1,3-Benzothiazole-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₅NOS
MW: 163.2 g/mol
Boiling Pt: 305.7 °C (760 mm Hg)
Melting Pt: 74 °C
Flash Pt: 138.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00526215
CAS Number: 6639-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR