1,3-benzothiazol-5-amine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,3-Benzothiazol-5-amine, 5-Aminobenzo[d]thiazole, 1,3-Benzothiazol-5-amine, 5-Aminobenzothiazole
A162547-100G
A162547-10G
A162547-250MG
A162547-5G
A162547-1G
A162547-25G
103805-624EA
15
USD
103805-624
103843-874
77011-272
77011-274
77011-276
77300-598
77319-920
77319-922
1,3-benzothiazol-5-amine 98%
1,3-benzothiazol-5-amine
Precaución: for Research Use Only
Formula:
C₇H₆N₂S MW: 150.199 g/mol Boiling Pt: 323.1 °C (760 mm Hg) Melting Pt: 55 °C Flash Pt: 149.2 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04115282 CAS Number: 1123-93-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...