1,3,6,8-Tetrabromopyrene 95%

Supplier: Ambeed
Warning

A428097-100G A428097-10G A428097-1G A428097-5G A428097-25G
103827-498EA 7 USD
103827-498 103838-554 76829-508 76829-510 76842-228 76842-230 77307-244
1,3,6,8-Tetrabromopyrene 95%
1,3,6,8-Tetrabromopyrene

Precaución: for Research Use Only

Formula: C₁₆H₆Br₄
MW: 517.84 g/mol
Boiling Pt: 548.2 °C (760 mm Hg)
Flash Pt: 273.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00428682
CAS Number: 128-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR