1,3,5-Tri-tert-butylbenzene 98%
Supplier: Ambeed
|
Warning
|
A212100-100G
A212100-10G
A212100-500G
A212100-1000G
A212100-25G
103809-472EA
407
USD
103809-472
103848-864
77192-668
77192-670
77192-672
77303-220
77303-222
1,3,5-Tri-tert-butylbenzene 98%
1,3,5-Tri-tert-butylbenzene
Precaución: for Research Use Only
Formula:
C₁₈H₃₀ Boiling Pt: 248 °C (760 mm Hg) Melting Pt: 67…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008831 CAS Number: 1460-02-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...