2,4,6-Tris(bromomethyl)mesitylene 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
1,3,5-Tris(bromomethyl)-2,4,6-trimethylbenzene, 2,4,6-Tris(bromomethyl)mesitylene
A275968-10G
A275968-25G
A275968-5G
103853-210EA
28
USD
103853-210
77194-056
77305-220
77305-222
2,4,6-Tris(bromomethyl)mesitylene 95%
2,4,6-Tris(bromomethyl)mesitylene
Precaución: for Research Use Only
Formula:
C₁₂H₁₅Br₃ Boiling Pt: 399.8 °C (760 mm Hg) Melting Pt: 187…189 °C Flash Pt: 189.6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00192554 CAS Number: 21988-87-4 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...