1,3,5-Trimethyl-1H-pyrazole 98%

Supplier: Ambeed
Danger

Synonyms: 1,3,5-Trimethyl-1H-pyrazole, 1,3,5-Trimethylpyrazole

A433627-5G A433627-10G A433627-25MG
103838-626EA 7 USD
103838-626 76842-328 77116-082 77116-084
1,3,5-Trimethyl-1H-pyrazole 98%
1,3,5-Trimethyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₆H₁₀N₂
MW: 110.16 g/mol
Melting Pt: 38…41 °C
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00015536
CAS Number: 1072-91-9
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR