1,3,5-Trifluoro-2-nitrobenzene 98%

Supplier: Ambeed
Warning

A152011-100G A152011-10G A152011-1G A152011-500G A152011-25G A152011-5G
103842-490EA 17 USD
103842-490 77286-148 77286-150 77286-152 77286-154 77318-090 77318-092
1,3,5-Trifluoro-2-nitrobenzene 98%
1,3,5-Trifluoro-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂F₃NO₂
Boiling Pt: 183.5 °C (760 mm Hg)
Melting Pt: 3.5 °C
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00014687
CAS Number: 315-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR