1,3,5-Trifluoro-2,4,6-triiodobenzene 98%

Supplier: Ambeed
Warning

A359516-1G A359516-25G A359516-5G A359516-100G A359516-100MG A359516-10G A359516-250MG
103826-230EA 15 USD
103826-230 103837-238 76840-412 76840-414 76840-416 77195-318 77195-320 77195-322 77195-324
1,3,5-Trifluoro-2,4,6-triiodobenzene 98%
1,3,5-Trifluoro-2,4,6-triiodobenzene

Precaución: for Research Use Only

Formula: C₆F₃I₃
MW: 509.77 g/mol
Boiling Pt: 352.4 °C (760 mm Hg)
Melting Pt: 152 °C
Flash Pt: 148.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06248899
CAS Number: 84322-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR