2,4,6-Tribromotoluene 97%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-2,4,6-tribromobenzene, 1,3,5-Tribromo-2-methylbenzene

A191700-1G A191700-25G A191700-100G A191700-10G A191700-5G
103816-394EA 9 USD
103816-394 77084-856 77084-858 77248-578 77248-580 77248-582
2,4,6-Tribromotoluene 97%
2,4,6-Tribromotoluene

Precaución: for Research Use Only

Formula: C₇H₅Br₃
MW: 328.83 g/mol
Boiling Pt: 290 °C (760 mm Hg)
Melting Pt: 67…70 °C
Flash Pt: 127.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00013527
CAS Number: 6320-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR