1,3,5-Tribromo-1,3,5-triazinane-2,4,6-trione 95%
Supplier: Ambeed
|
Danger
|
A300302-250MG
A300302-1G
103814-268EA
15
USD
103814-268
103835-420
76816-758
77305-804
1,3,5-Tribromo-1,3,5-triazinane-2,4,6-trione 95%
1,3,5-Tribromo-1,3,5-triazinane-2,4,6-trione
Precaución: for Research Use Only
Formula:
C₃Br₃N₃O₃ Boiling Pt: 324.3±25.0 °C (760 mm Hg) Flash Pt: 149.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD21604032 CAS Number: 17497-85-7 UN: 3085 ADR: 5.1,8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...