1,3,4-oxadiazol-2-amine 95%

Supplier: Ambeed
Warning

Synonyms: 1,3,4-Oxadiazol-2-amine

A111979-10G A111979-250MG A111979-1G A111979-5G
103797-930EA 29 USD
103797-930 103823-754 76970-882 76970-884 77310-562 77310-564
1,3,4-oxadiazol-2-amine 95%
1,3,4-oxadiazol-2-amine

Precaución: for Research Use Only

Formula: C₂H₃N₃O
MW: 85.065 g/mol
Boiling Pt: 205.6 °C (760 mm Hg)
Flash Pt: 78.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD17677516
CAS Number: 3775-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR