1,3,3-Trimethylindolinonaphthospirooxazine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,3,3-Trimethylspiro[indoline-2,3'-[3H]naphth[2,1-b][1,4]oxazine], 1,3,3-Trimethylspiro[indoline-2,3'-naphtho[2,1-b][1,4]oxazine]
A254660-100MG
A254660-10G
A254660-250MG
A254660-1G
A254660-5G
103812-148EA
26
USD
103812-148
103852-002
77000-758
77000-760
77000-762
77304-636
77304-638
1,3,3-Trimethylindolinonaphthospirooxazine 98%
1,3,3-Trimethylindolinonaphthospirooxazine
Precaución: for Research Use Only
Formula:
C₂₂H₂₀N₂O MW: 328.42 g/mol Boiling Pt: 529.1 °C (760 mm Hg) Melting Pt: 126…128 °C Flash Pt: 273.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013318 CAS Number: 27333-47-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...