1,3,2-Dioxathiolane-2,2-dioxide 98%

Supplier: Ambeed
Warning

A265284-100G A265284-10G A265284-500G A265284-5G A265284-25G
103852-612EA 5 USD
103852-612 76801-868 76801-870 76801-872 77092-284 77304-914
1,3,2-Dioxathiolane-2,2-dioxide 98%
1,3,2-Dioxathiolane-2,2-dioxide

Precaución: for Research Use Only

Formula: C₂H₄O₄S
Boiling Pt: 231.1 °C (760 mm Hg)
Melting Pt: 95…97 °C
Flash Pt: 93.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00221769
CAS Number: 1072-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR