(±)-1,2-Pentanediol 98%

Supplier: Ambeed
Warning

Synonyms: (±)-1,2-Dihydroxy pentane, 1,2-Pentanediol

A101958-100G A101958-25G A101958-500G A101958-1G A101958-5G
103796-228EA 5 USD
103796-228 103821-922 77188-572 77188-574 77188-576 77282-876 77308-808
(±)-1,2-Pentanediol 98%
(±)-1,2-Pentanediol

Precaución: for Research Use Only

Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 206 °C (760 mm Hg)
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00010736
CAS Number: 5343-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR