Deoxybenzoin 98%

Supplier: Ambeed
Warning

A246355-100G A246355-10G A246355-1000G A246355-25G A246355-5G A246355-500G
103809-758EA 191 USD
103809-758 103851-536 76692-368 76692-370 77090-830 77090-832 77304-386 77877-825
Deoxybenzoin 98%
Deoxybenzoin

Precaución: for Research Use Only

Formula: C₁₄H₁₂O
Boiling Pt: 319…321 °C (760 mm Hg)
Melting Pt: 53…57 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003081
CAS Number: 451-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR