1,2-Diphenylethane 98%

Supplier: Ambeed
Warning

Synonyms: Dibenzyl, Bibenzyl

A695765-1000G A695765-100G A695765-500G A695765-25G
103840-604EA 5 USD
103840-604 76697-278 76697-280 76697-282 76844-824
1,2-Diphenylethane 98%
1,2-Diphenylethane

Precaución: for Research Use Only

Formula: (C₆H₅)CH₂CH₂(C₆H₅)
MW: 182.27 g/mol
Boiling Pt: 280.5 °C (760 mm Hg)
Melting Pt: 49…53 °C (760 mm Hg)
Flash Pt: 106.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00004796
CAS Number: 103-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR