1,2-Dimethyl-1,4,5,6-tetrahydropyrimidine 97%

Supplier: Ambeed
Warning

A217088-25G A217088-500G
103809-526EA 283 USD
103809-526 103849-326 77087-938 77087-940 77192-804 77192-806
1,2-Dimethyl-1,4,5,6-tetrahydropyrimidine 97%
1,2-Dimethyl-1,4,5,6-tetrahydropyrimidine

Precaución: for Research Use Only

Formula: C₆H₁₂N₂
Boiling Pt: 191.7 °C (760 mm Hg)
Flash Pt: 69.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036056
CAS Number: 4271-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR