1,2-Diiodobenzene 98% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilized with Copper chip

A191924-1G A191924-1000G A191924-100G A191924-10G A191924-500G A191924-25G A191924-5G
103816-410EA 5 USD
103816-410 103847-012 77084-880 77111-802 77111-804 77111-806 77111-808 77302-274 77302-276
1,2-Diiodobenzene 98% stabilized
1,2-Diiodobenzene

Precaución: for Research Use Only

Formula: C₆H₄I₂
MW: 329.91 g/mol
Boiling Pt: 284.8 °C (760 mm Hg)
Melting Pt: 23…27 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001038
CAS Number: 615-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR