1,2-Diethoxybenzene 98%

Supplier: Ambeed
Warning

A105762-100G A105762-25G A105762-5G
103822-676EA 6 USD
103822-676 77107-578 77107-580 77283-392
1,2-Diethoxybenzene 98%
1,2-Diethoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Boiling Pt: 218.7 °C (760 mm Hg)
Melting Pt: 43…45 °C
Flash Pt: 73.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00015143
CAS Number: 2050-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR