1,2-Dibromotetrachloroethane 97%

Supplier: Ambeed
Warning

A642292-25G A642292-5G A642292-100G A642292-500G A642292-1G
103840-346EA 6 USD
103840-346 76844-522 76844-524 77198-388 77198-390 77308-044
1,2-Dibromotetrachloroethane 97%
1,2-Dibromotetrachloroethane

Precaución: for Research Use Only

Formula: BrCl₂CCBrCl₂
MW: 325.64 g/mol
Melting Pt: 215…220 °C
Flash Pt: 104.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00000780
CAS Number: 630-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR