1,2-Dibromo-4-nitrobenzene 97%

Supplier: Ambeed
Warning

A102042-10G A102042-250MG A102042-25G A102042-1G A102042-5G
103796-260EA 6 USD
103796-260 103821-958 76982-198 76982-200 76982-202 77308-844 77308-846
1,2-Dibromo-4-nitrobenzene 97%
1,2-Dibromo-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃Br₂NO₂
Melting Pt: 53…55 °C
Storage Temperature: Ambient
MDL Number: MFCD00055373
CAS Number: 5411-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR