3,4-Dibromotoluene 98%

Supplier: Ambeed

A163712-100G A163712-10G A163712-25G
103844-016EA 7 USD
103844-016 77110-764 77110-766 77320-124 77320-126
3,4-Dibromotoluene 98%
3,4-Dibromotoluene

Precaución: for Research Use Only

Formula: C₇H₆Br₂
MDL Number: MFCD00079744
CAS Number: 60956-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR