4,5-Dibromo-o-xylene 98%

Supplier: Ambeed
Warning

Synonyms: 1,2-Dibromo-4,5-dimethylbenzene, 4,5-Dibromo-1,2-dimethylbenzene

A226942-100G A226942-10G A226942-500G A226942-1G A226942-25G A226942-5G
103819-280EA 5 USD
103819-280 103850-188 76691-956 76691-958 76691-960 77088-936 77088-938 77088-940
4,5-Dibromo-o-xylene 98%
4,5-Dibromo-o-xylene

Precaución: for Research Use Only

Formula: C₈H₈Br₂
Boiling Pt: 279.1 °C (760 mm Hg)
Flash Pt: 138.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00082743
CAS Number: 24932-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR