4,5-Dibromoveratrole 95%

Supplier: Ambeed
Warning

Synonyms: 1,2-Dibromo-4,5-dimethoxybenzene, 4,5-Dibromoveratrole

A121263-100G A121263-25G A121263-5G
77284-810EA 169.95 USD
77284-810 77284-812 77312-168
4,5-Dibromoveratrole 95%
4,5-Dibromoveratrole

Precaución: for Research Use Only

Formula: C₈H₈Br₂O₂
MW: 295.96 g/mol
Boiling Pt: 286.6 °C (760 mm Hg)
Flash Pt: 114.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00014894
CAS Number: 37895-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR