1,2-Dibromo-4,5-difluorobenzene 98%

Supplier: Ambeed
Danger

A109867-5G A109867-25G
103823-388EA 5 USD
103823-388 77310-194 77549-124
1,2-Dibromo-4,5-difluorobenzene 98%
1,2-Dibromo-4,5-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂F₂
Melting Pt: 34…35 °C
Storage Temperature: Ambient
MDL Number: MFCD00009890
CAS Number: 64695-78-9
UN: 1993
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR