1,2-Diamino-3-chlorobenzene 97%

Supplier: Ambeed
Warning

Synonyms: 3-Chlorobenzene-1,2-diamine

A111572-10G A111572-250MG A111572-25G A111572-1G A111572-5G
103797-864EA 12 USD
103797-864 103823-682 76970-822 76970-824 76970-826 77310-494 77310-496
1,2-Diamino-3-chlorobenzene 97%
1,2-Diamino-3-chlorobenzene

Precaución: for Research Use Only

Formula: C₆H₇ClN₂
MW: 142.59 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09881742
CAS Number: 21745-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR