1,2-Di(4-pyridyl)ethane 95%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Ethylenedipyridine, 1,2-Di(pyridin-4-yl)ethane, 1,2-Bis(4-pyridyl)ethane

A697029-1G A697029-5G A697029-10G
103840-612EA 58 USD
103840-612 76844-832 76844-834 77198-798
1,2-Di(4-pyridyl)ethane 95%
1,2-Di(4-pyridyl)ethane

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂
MW: 184.24 g/mol
Boiling Pt: 310.3 °C (760 mm Hg)
Melting Pt: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00006451
CAS Number: 4916-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR