2,2'-Dipyridyl disulfide 98%

Supplier: Ambeed
Warning

Synonyms: 1,2-Di(pyridin-2-yl)disulphane, 2,2'-Dithiodipyridine

A172843-25G A172843-1000G A172843-100G A172843-500G A172843-5G
103845-046EA 8 USD
103845-046 77082-260 77111-076 77111-078 77111-080 77301-222
2,2'-Dipyridyl disulfide 98%
2,2'-Dipyridyl disulphide

Precaución: for Research Use Only

Formula: C₁₀H₈N₂S₂
MW: 220.32 g/mol
Boiling Pt: 356 °C (760 mm Hg)
Melting Pt: 56…58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006287
CAS Number: 2127-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR