1,2-Cyclohexanediol 95%

Supplier: Ambeed
Warning

Synonyms: Cyclohexane-1,2-diol

A716626-25G A716626-5G A716626-100G A716626-10G A716626-500G
103840-714EA 7 USD
103840-714 76844-928 76844-930 77198-962 77198-964 77198-966
1,2-Cyclohexanediol 95%
1,2-Cyclohexanediol

Precaución: for Research Use Only

Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 118…120 °C (10 mm Hg)
Melting Pt: 73…77 °C
Flash Pt: 114.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00003861
CAS Number: 931-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR