1,2-Bis(diphenylphosphino)ethane 97%

Supplier: Ambeed
Warning

Synonyms: Ethylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)ethane, DIPHOS, 1,2-Bis(diphenylphosphino)ethane, Ethane-1,3-diylbis(diphenylphosphine)

A103846-100G A103846-10G A103846-25G A103846-500G A103846-5G
103822-326EA 5 USD
103822-326 76994-684 76994-686 76994-688 76994-690 77309-192
1,2-Bis(diphenylphosphino)ethane 97%
1,2-Bis(diphenylphosphino)ethane

Precaución: for Research Use Only

Formula: C₂₆H₂₄P₂
Boiling Pt: 514.8 °C (760 mm Hg)
Melting Pt: 137…142 °C
Flash Pt: 281.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00003047
CAS Number: 1663-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR