1,2-Bis(diphenylphosphino)benzene 97%
Supplier: Ambeed
|
Warning
|
A149760-10G
A149760-250MG
A149760-1G
A149760-25G
A149760-5G
A149760-100G
103804-076EA
18
USD
103804-076
103842-202
76997-286
76997-288
77317-704
77317-706
77317-708
77881-167
1,2-Bis(diphenylphosphino)benzene 97%
1,2-Bis(diphenylphosphino)benzene
Precaución: for Research Use Only
Formula:
C₃₀H₂₄P₂ Boiling Pt: 546.9 °C (760 mm Hg) Melting Pt: 185…187 °C Flash Pt: 302.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014081 CAS Number: 13991-08-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...