1,2-Bis(dimethylsilyl)benzene 97%

Supplier: Ambeed
Warning

Synonyms: 1,2-Phenylenebis(dimethylsilane)

A127565-10G A127565-1G
103800-984EA 18 USD
103800-984 77189-962 77298-228
1,2-Bis(dimethylsilyl)benzene 97%
1,2-Bis(dimethylsilyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₁₈Si₂
Boiling Pt: 220 °C (760 mm Hg)
Melting Pt: <25 °C
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00142462
CAS Number: 17985-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR