1,2-Bis(bromomethyl)-4-nitrobenzene 97%

Supplier: Ambeed
Danger

A102048-250MG A102048-5G
103796-268EA 236 USD
103796-268 103821-964 77188-584 77308-848
1,2-Bis(bromomethyl)-4-nitrobenzene 97%
1,2-Bis(bromomethyl)-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₇Br₂NO₂
Boiling Pt: 384.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08234715
CAS Number: 6425-66-7
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR