Bis(4-bromophenyl)acetylene 97%

Supplier: Ambeed
Warning

A310485-100G A310485-10G A310485-250MG A310485-25G A310485-5G A310485-1G
103814-624EA 38 USD
103814-624 103835-806 76693-548 76693-550 76693-552 76838-580 76838-582 77306-024
Bis(4-bromophenyl)acetylene 97%
Bis(4-bromophenyl)acetylene

Precaución: for Research Use Only

Formula: C₁₄H₈Br₂
Boiling Pt: 383.6 °C (760 mm Hg)
Melting Pt: 182 °C
Storage Temperature: Ambient
MDL Number: MFCD00185147
CAS Number: 2789-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR