2-(Hydroxymethyl)benzyl alcohol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
o-Phenylenedimethanol, o-Xylene-α,α'-diol, 1,2-Benzenedimethanol
A123290-100G
A123290-25G
A123290-500G
A123290-1G
A123290-5G
103800-380EA
14
USD
103800-380
77189-748
77189-750
77189-752
77297-938
77312-500
2-(Hydroxymethyl)benzyl alcohol 98%
2-(Hydroxymethyl)benzyl alcohol
Precaución: for Research Use Only
Formula:
C₈H₁₀O₂ MW: 138.17 g/mol Boiling Pt: 291 °C (760 mm Hg) Melting Pt: 59…64 °C Flash Pt: 145 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004626 CAS Number: 612-14-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...