1,2:5,6-Di-O-isopropylidene-α-D-allofuranose 97%

Supplier: Ambeed
Warning

Synonyms: 1,2:5,6-Di-o-isopropylidene-alpha-D-allofuranose

A486116-1G A486116-25G A486116-100G A486116-10G A486116-500G
103828-178EA 7 USD
103828-178 103839-252 76843-192 76843-194 77196-836 77196-838 77196-840
1,2:5,6-Di-O-isopropylidene-α-D-allofuranose 97%
1,2:5,6-Di-O-isopropylidene-alpha-D-allofuranose

Precaución: for Research Use Only

Formula: C₁₂H₂₀O₆
MW: 260.28 g/mol
Boiling Pt: 362.8 °C (760 mm Hg)
Melting Pt: 73…76 °C
Flash Pt: 173.2 °C
Storage Temperature: Freezer
MDL Number: MFCD00135634
CAS Number: 2595-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR