6-Azauracil 98%

Supplier: Ambeed
Warning

Synonyms: 1,2,4-Triazine-3,5(2H,4H)-dione

A238332-1G A238332-5G A238332-100G A238332-10G A238332-25G A238332-500G
103811-220EA 8 USD
103811-220 103851-000 77090-112 77090-114 77275-944 77275-946 77275-948 77275-950
6-Azauracil 98%
6-Azauracil

Precaución: for Research Use Only

Formula: C₃H₃N₃O₂
MW: 113.08 g/mol
Melting Pt: 274…275 °C
Storage Temperature: Ambient
MDL Number: MFCD00006456
CAS Number: 461-89-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR