1,2,3-Trifluoro-4-nitrobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 2,3,4-Trifluoronitrobenzene

A210978-1000G A210978-25G A210978-100G A210978-10G A210978-500G
103809-460EA 107 USD
103809-460 77087-142 77087-144 77287-942 77287-944 77287-946
1,2,3-Trifluoro-4-nitrobenzene 98%
1,2,3-Trifluoro-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂F₃NO₂
MW: 177.08 g/mol
Boiling Pt: 92 °C (20 mm Hg)
Flash Pt: 63.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00041546
CAS Number: 771-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR