1,2,3-Trichloro-5-nitrobenzene 97%

Supplier: Ambeed
Warning

Synonyms: 1-Nitro-3,4,5-trichlorobenzene, 5-Nitro-1,2,3-trichlorobenzene

A170159-25G A170159-5G A170159-100G A170159-10G A170159-500G
103806-454EA 6 USD
103806-454 103844-720 77081-824 77081-826 77096-478 77096-480 77096-482
1,2,3-Trichloro-5-nitrobenzene 97%
1,2,3-Trichloro-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂Cl₃NO₂
Boiling Pt: 284.1 °C (760 mm Hg)
Melting Pt: 68…71 °C
Flash Pt: 125.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00061096
CAS Number: 20098-48-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR